2-Amino-5-brombenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Amino-5-Bromo-Benzenemethanol, (2-Amino-5-bromophenyl)methanol, (2-Amino-5-Bromophenyl)methonal

AS35749G1 AS35749G5
ACTIAS35749G1EA 0 CHF
ACTIAS35749G1 ACTIAS35749G5
2-Amino-5-brombenzylalkohol
2-Amino-5-brombenzylalkohol
Formel: C₇H₈BrNO
MW: 202,0500 g/mol
MDL Number: MFCD08234509
CAS-Nummer: 20712-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR