4-Phenyl-3-butin-2-on

Lieferant: ACTIVATE SCIENTIFIC

AS35405G1 AS35405G5
ACTIAS35405G1EA 0 CHF
ACTIAS35405G1 ACTIAS35405G5
4-Phenyl-3-butin-2-on
4-Phenyl-3-butin-2-on
Formel: C₁₀H₈O
MW: 144,17 g/mol
MDL Number: MFCD00008776
CAS-Nummer: 1817-57-8
EINECS: 217-327-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR