Thiomorpholin-1,1-dioxid Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Thiomorpholine 1,1-dioxide hydrochloride

AS15190G1 AS15190G5
ACTIAS15190G1EA 0 CHF
ACTIAS15190G1 ACTIAS15190G5
Thiomorpholin-1,1-dioxid Hydrochlorid
Thiomorpholin-1,1-dioxid Hydrochlorid
Formel: C₄H₁₀ClNO₂S
MW: 171,6457 g/mol
MDL Number: MFCD06738995
CAS-Nummer: 59801-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR