1,2,4,5-Tetrakis(tert-butylthio)benzol

Lieferant: ACTIVATE SCIENTIFIC

AS35489G1 AS35489G5
ACTIAS35489G1EA 0 CHF
ACTIAS35489G1 ACTIAS35489G5
1,2,4,5-Tetrakis(tert-butylthio)benzol
1,2,4,5-Tetrakis(tert-butylthio)benzol
Formel: C₂₂H₃₈S₄
MDL Number: MFCD08276431
CAS-Nummer: 447463-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR