3-(Methylsulfonyl)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS22329G1 AS22329G25 AS22329G5
ACTIAS22329G1EA 59 CHF
ACTIAS22329G1 ACTIAS22329G25 ACTIAS22329G5
3-(Methylsulfonyl)anilin
3-(Methylsulfonyl)anilin
Formel: C₇H₉NO₂S
MDL Number: MFCD03764428
CAS-Nummer: 35216-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR