4-(Methylsulfonyl)-2-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS20444G1 AS20444G5
ACTIAS20444G1EA 0 CHF
ACTIAS20444G1 ACTIAS20444G5
4-(Methylsulfonyl)-2-nitroanilin
4-(Methylsulfonyl)-2-nitroanilin
Formel: C₇H₈N₂O₄S
MDL Number: MFCD00718367
CAS-Nummer: 21731-56-6
EINECS: 244-550-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR