N-Boc-3-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-N-Boc-3-Fluoroaniline, 3-Fluoroaniline, N-BOC protected

AS35939G1 AS35939G5
ACTIAS35939G1EA 0 CHF
ACTIAS35939G1 ACTIAS35939G5
N-Boc-3-fluoranilin
N-Boc-3-fluoranilin
Formel: C₁₁H₁₄FNO₂
MW: 211.2328 g/mol
MDL Number: MFCD04035656
CAS-Nummer: 81740-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR