N-Boc-4-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: tert-Butyl (4-nitrophenyl)carbamate

AS13056G1 AS13056G5
ACTIAS13056G1EA 0 CHF
ACTIAS13056G1 ACTIAS13056G5
N-Boc-4-nitroanilin
N-Boc-4-nitroanilin
Formel: C₁₁H₁₄N₂O₄
MW: 238,2399 g/mol
MDL Number: MFCD01006654
CAS-Nummer: 18437-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR