3-(Methylsulfonyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS20770G1 AS20770G25 AS20770G5
ACTIAS20770G1EA 0 CHF
ACTIAS20770G1 ACTIAS20770G25 ACTIAS20770G5
3-(Methylsulfonyl)benzonitril
3-(Methylsulfonyl)benzonitril
Formel: C₈H₇NO₂S
MDL Number: MFCD04037969
CAS-Nummer: 22821-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR