2-(Methylsulfonyl)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS20778G25 AS20778G5
ACTIAS20778G25EA 858 CHF
ACTIAS20778G25 ACTIAS20778G5
2-(Methylsulfonyl)benzonitril
2-(Methylsulfonyl)benzonitril
Formel: C₈H₇NO₂S
MDL Number: MFCD11052333
CAS-Nummer: 89942-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR