2-Chlor-5-cyanphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51177G1 AS51177G5
ACTIAS51177G1EA 0 CHF
ACTIAS51177G1 ACTIAS51177G5
2-Chlor-5-cyanphenylboronsäure
2-Chlor-5-cyanphenylboronsäure
Formel: C₇H₅BClNO₂
MW: 181.3841 g/mol
MDL Number: MFCD06656271
CAS-Nummer: 936249-33-1

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR