N-Boc-L-alaninol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol

AS25057G25 AS25057G5
ACTIAS25057G25EA 40 CHF
ACTIAS25057G25 ACTIAS25057G5
N-Boc-L-alaninol
N-Boc-L-alaninol
Formel: C₈H₁₇NO₃
MW: 175,2 g/mol
MDL Number: MFCD00043121
CAS-Nummer: 79069-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR