3-(Methansulfonylamino)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS20868G1 AS20868G5
ACTIAS20868G1EA 0 CHF
ACTIAS20868G1 ACTIAS20868G5
3-(Methansulfonylamino)benzonitril
3-(Methansulfonylamino)benzonitril
Formel: C₈H₈N₂O₂S
MDL Number: MFCD00116199
CAS-Nummer: 50790-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR