2-(Methansulfonylamino)benzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS20869G1 AS20869G5
ACTIAS20869G1EA 133 CHF
ACTIAS20869G1 ACTIAS20869G5
2-(Methansulfonylamino)benzonitril
2-(Methansulfonylamino)benzonitril
Formel: C₈H₈N₂O₂S
MDL Number: MFCD00590384
CAS-Nummer: 50790-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR