Picolinamidin Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

AS1234G1 AS1234G5
ACTIAS1234G1EA 0 CHF
ACTIAS1234G1 ACTIAS1234G5
Picolinamidin Hydrochlorid
Picolinamidin Hydrochlorid
Formel: C₆H₈ClN₃
MDL Number: MFCD00052271
CAS-Nummer: 51285-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR