Ethyl-2,4-dimethylpyrrol-3-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate

AS30631G1 AS30631G5
ACTIAS30631G1EA 0 CHF
ACTIAS30631G1 ACTIAS30631G5
Ethyl-2,4-dimethylpyrrol-3-carboxylat
Ethyl-2,4-dimethylpyrrol-3-carboxylat
Formel: C₉H₁₃NO₂
MW: 167,2050 g/mol
MDL Number: MFCD01365811
CAS-Nummer: 2199-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR