Ethyl-4-amino-2-(methylthio)pyrimidin-5-carboxylat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 4-amino-2-(methylthio)pyrimidin-5-carboxylate, Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate

AS1398G1 AS1398G5
ACTIAS1398G1EA 0 CHF
ACTIAS1398G1 ACTIAS1398G5
Ethyl-4-amino-2-(methylthio)pyrimidin-5-carboxylat
Ethyl 4-amino-2-(methylthio)pyrimidine-5-carboxylate
Formel: C₈H₁₁N₃O₂S
MW: 213,2568 g/mol
MDL Number: MFCD00090781
CAS-Nummer: 776-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR