2-(Methylthio)-5-pyrimidinboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(2-(Methylthio)pyrimidin-5-yl)boronic acid, 2-(Methylthio)pyrimidine-5-boronic acid
AS51170G1
AS51170G5
ACTIAS51170G1EA
0
CHF
ACTIAS51170G1
ACTIAS51170G5
2-(Methylthio)-5-pyrimidinboronsäure
2-(Methylthio)-5-pyrimidineboronic acid
Formel:
C₅H₇BN₂O₂S MW: 170.0000 g/mol |
MDL Number:
MFCD07375142 CAS-Nummer: 348098-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...