(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: N-Boc-1H-pyrazole-4-boronic acid, 1-tert-Butoxycarbonyl-1H-pyrazole-4-boronic acid, 1H-Pyrazole-4-boronic acid, N1-BOC protected

AS32622G1 AS32622G5
ACTIAS32622G1EA 0 CHF
ACTIAS32622G1 ACTIAS32622G5
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid
(1-(tert-Butoxycarbonyl)-1H-pyrazol-4-yl)boronic acid
Formel: C₈H₁₃BN₂O₄
MW: 212.01 g/mol
MDL Number: MFCD09951915
CAS-Nummer: 947533-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR