1-Methyl-4-imidazolsulfonylchlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Methyl-1H-imidazole-4-sulphonyl chloride

AS21756G100 AS21756G25 AS21756G5
ACTIAS21756G100EA 0 CHF
ACTIAS21756G100 ACTIAS21756G25 ACTIAS21756G5
1-Methyl-4-imidazolsulfonylchlorid
1-Methyl-4-imidazolsulfonylchlorid
Formel: C₄H₅ClN₂O₂S
MDL Number: MFCD00068060
CAS-Nummer: 137049-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR