3,4-(Ethylendioxy)benzylamin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-(Aminomethyl)-2,3-dihydro-1,4-benzodioxine, (2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)methanamine

AS51335G1 AS51335G5
ACTIAS51335G1EA 0 CHF
ACTIAS51335G1 ACTIAS51335G5
3,4-(Ethylendioxy)benzylamin
3,4-(Ethylendioxy)benzylamin
Formel: C₈H₉NO₂
MDL Number: MFCD02677684
CAS-Nummer: 17413-10-4
EINECS: 241-440-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR