6-(Trifluormethyl)isatin

Lieferant: ACTIVATE SCIENTIFIC

AS35802G1 AS35802G10 AS35802G5
ACTIAS35802G1EA 0 CHF
ACTIAS35802G1 ACTIAS35802G10 ACTIAS35802G5
6-(Trifluormethyl)isatin
6-(Trifluormethyl)isatin
Formel: C₉H₄F₃NO₂
MDL Number: MFCD08457887
CAS-Nummer: 343-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR