6-(Trifluormethoxy)isatin

Lieferant: ACTIVATE SCIENTIFIC

AS36594G1 AS36594G5
ACTIAS36594G1EA 0 CHF
ACTIAS36594G1 ACTIAS36594G5
6-(Trifluormethoxy)isatin
6-(Trifluormethoxy)isatin
Formel: C₉H₄F₃NO₃
MDL Number: MFCD06804530
CAS-Nummer: 162252-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR