4-(4-Methoxybenzoyl)piperidin Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

AS36726G1 AS36726G5
ACTIAS36726G1EA 0 CHF
ACTIAS36726G1 ACTIAS36726G5
4-(4-Methoxybenzoyl)piperidin Hydrochlorid
4-(4-Methoxybenzoyl)piperidin Hydrochlorid
Formel: C₁₃H₁₈ClNO₂
MDL Number: MFCD00114817
CAS-Nummer: 25519-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR