N(α)-Boc-L-tryptophan
Lieferant: BACHEM BIOCHEMICA
Synonyms:
Boc-Trp-OH
BACE4000240.0500EA
449
CHF
BACE4000240.0500
BACE4000240.1000
BACE4000240.0100
N(α)-Boc-L-tryptophan
N(α)-Boc-L-tryptophan
Formel:
C₁₆H₂₀N₂O₄ MW: 304,3 g/mol |
MDL Number:
MFCD00065595 CAS-Nummer: 13139-14-5 EINECS: 236-072-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...