1-Boc-2-ethylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: tert-Butyl 2-ethylpiperazine-1-carboxylate

AS31944G1 AS31944G5
ACTIAS31944G1EA 0 CHF
ACTIAS31944G1 ACTIAS31944G5
1-Boc-2-ethylpiperazin
1-Boc-2-ethylpiperazin
Formel: C₁₁H₂₂N₂O₂
MW: 214,3100 g/mol
MDL Number: MFCD03265490
CAS-Nummer: 393781-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR