(S)-1-Boc-3-ethylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (S)-tert-Butyl 3-ethylpiperazine-1-carboxylate

AS3125G5 AS3125G1 AS3125G25
ACTIAS3125G5EA 0 CHF
ACTIAS3125G5 ACTIAS3125G1 ACTIAS3125G25
(S)-1-Boc-3-ethylpiperazin
(S)-1-Boc-3-ethylpiperazin
Formel: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
MDL Number: MFCD07772110
CAS-Nummer: 928025-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR