(S)-1-Boc-3-isobutylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

AS3139G1 AS3139G5
ACTIAS3139G1EA 0 CHF
ACTIAS3139G1 ACTIAS3139G5
(S)-1-Boc-3-isobutylpiperazin
(S)-1-Boc-3-isobutylpiperazin
Formel: C₁₃H₂₆N₂O₂
MDL Number: MFCD07772108
CAS-Nummer: 928025-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR