(S)-1-Boc-2-ethylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (2S)-2-Ethylpiperazine, N1-BOC protected, (S)-tert-Butyl 2-ethylpiperazine-1-carboxylate

AS3120G1 AS3120G5
ACTIAS3120G1EA 0 CHF
ACTIAS3120G1 ACTIAS3120G5
(S)-1-Boc-2-ethylpiperazin
(S)-1-Boc-2-ethylpiperazin
Formel: C₁₁H₂₂N₂O₂
MW: 214,30 g/mol
MDL Number: MFCD07772100
CAS-Nummer: 325145-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR