(R)-1-Boc-3-propylpiperazin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
tert-Butyl (R)-3-propylpiperazine-1-carboxylate
AS3128G1
AS3128G5
ACTIAS3128G1EA
0
CHF
ACTIAS3128G1
ACTIAS3128G5
(R)-1-Boc-3-propylpiperazin
(R)-1-Boc-3-propylpiperazin
Formel:
C₁₂H₂₄N₂O₂ MW: 228,3312 g/mol |
MDL Number:
MFCD07772107 CAS-Nummer: 928025-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...