(R)-(+)-1-Boc-3-methylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

AS31896G1 AS31896G5
ACTIAS31896G1EA 0 CHF
ACTIAS31896G1 ACTIAS31896G5
(R)-(+)-1-Boc-3-methylpiperazin
(R)-(+)-1-Boc-3-methylpiperazin
Formel: C₁₀H₂₀N₂O₂
MDL Number: MFCD02683205
CAS-Nummer: 163765-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR