(R)-1-Boc-3-isopropylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

AS3130G1 AS3130G5
ACTIAS3130G1EA 0 CHF
ACTIAS3130G1 ACTIAS3130G5
(R)-1-Boc-3-isopropylpiperazin
(R)-1-Boc-3-isopropylpiperazin
Formel: C₁₂H₂₄N₂O₂
MW: 228.3300 g/mol
MDL Number: MFCD07772113
CAS-Nummer: 928025-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR