(R)-1-Boc-2-ethylpiperazin

Lieferant: ACTIVATE SCIENTIFIC

AS3119G1 AS3119G5
ACTIAS3119G1EA 0 CHF
ACTIAS3119G1 ACTIAS3119G5
(R)-1-Boc-2-ethylpiperazin
(R)-1-Boc-2-ethylpiperazin
Formel: C₁₁H₂₂N₂O₂
MW: 214,3100 g/mol
MDL Number: MFCD03411920
CAS-Nummer: 393781-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR