4-(Morpholinosulfonyl)anilin

Lieferant: ACTIVATE SCIENTIFIC

AS35345G1 AS35345G5
ACTIAS35345G1EA 0 CHF
ACTIAS35345G1 ACTIAS35345G5
4-(Morpholinosulfonyl)anilin
4-(Morpholinosulfonyl)anilin
Formel: C₁₀H₁₄N₂O₃S
MDL Number: MFCD00277213
CAS-Nummer: 21626-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR