1-Boc-piperidin-4-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

AS20295G100 AS20295G25
ACTIAS20295G100EA 0 CHF
ACTIAS20295G100 ACTIAS20295G25
1-Boc-piperidin-4-carbonsäure
1-Boc-piperidin-4-carbonsäure
Formel: C₁₁H₁₉NO₄
MW: 229,3 g/mol
MDL Number: MFCD00076999
CAS-Nummer: 84358-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR