(S)-1-Boc-piperidin-3-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (S)-Boc-nipecotic acid, 1-Boc-L-nipecotic acid, (S)-1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid

AS31713G1 AS31713G5
ACTIAS31713G1EA 21 CHF
ACTIAS31713G1 ACTIAS31713G5
(S)-1-Boc-piperidin-3-carbonsäure
(S)-1-Boc-piperidin-3-carbonsäure
Formel: C₁₁H₁₉NO₄
MW: 229,28 g/mol
MDL Number: MFCD02179172
CAS-Nummer: 88495-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR