1-Boc-3-cyanpiperidin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS21277G1EA 0 CHF
ACTIAS21277G1 ACTIAS21277G5
1-Boc-3-cyanpiperidin
1-Boc-3-cyanpiperidin
Formel: C₁₁H₁₈N₂O₂
MW: 210,27 g/mol
MDL Number: MFCD01861222
CAS-Nummer: 91419-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR