1-Boc-2-cyanpiperidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: tert-Butyl 2-cyanopiperidine-1-carboxylate

AS40068G1 AS40068G5
ACTIAS40068G1EA 0 CHF
ACTIAS40068G1 ACTIAS40068G5
1-Boc-2-cyanpiperidin
1-Boc-2-cyanpiperidin
Formel: C₁₁H₁₈N₂O₂
MW: 210,27 g/mol
MDL Number: MFCD01861221
CAS-Nummer: 153749-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR