(R)-Ethyl-3-piperidincarboxylat Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (R)-Ethyl piperidine-3-carboxylate hydrochloride

AS35936G1 AS35936G5
ACTIAS35936G1EA 0 CHF
ACTIAS35936G1 ACTIAS35936G5
(R)-Ethyl-3-piperidincarboxylat Hydrochlorid
(R)-Ethyl-3-piperidincarboxylat Hydrochlorid
Formel: C₈H₁₆ClNO₂
MW: 193.6711 g/mol
MDL Number: MFCD05864623
CAS-Nummer: 37675-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR