1-(Dimethylcarbamoyl)piperidin-2-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS23959G1 AS23959G5
ACTIAS23959G1EA 0 CHF
ACTIAS23959G1 ACTIAS23959G5
1-(Dimethylcarbamoyl)piperidin-2-carbonsäure
1-(Dimethylcarbamoyl)piperidin-2-carbonsäure
Formel: C₉H₁₆N₂O₃
MDL Number: MFCD14596111
CAS-Nummer: 1249531-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR