(1-Trityl-1H-imidazol-4-yl)methanol

Lieferant: ACTIVATE SCIENTIFIC

AS50047G1 AS50047G5
ACTIAS50047G1EA 0 CHF
ACTIAS50047G1 ACTIAS50047G5
(1-Trityl-1H-imidazol-4-yl)methanol
(1-Trityl-1H-imidazol-4-yl)methanol
Formel: C₂₃H₂₀N₂O
MDL Number: MFCD02179553
CAS-Nummer: 33769-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR