4-(Chlormethyl)-1-tritylimidazol

Lieferant: ACTIVATE SCIENTIFIC

AS50283G1 AS50283G5
ACTIAS50283G1EA 0 CHF
ACTIAS50283G1 ACTIAS50283G5
4-(Chlormethyl)-1-tritylimidazol
4-(Chlormethyl)-1-tritylimidazol
Formel: C₂₃H₁₉ClN₂
MDL Number: MFCD06660115
CAS-Nummer: 103057-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR