1-Methyl-2-(tributylstannyl)indol

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51235G1 AS51235G5
ACTIAS51235G1EA 0 CHF
ACTIAS51235G1 ACTIAS51235G5
1-Methyl-2-(tributylstannyl)indol
1-Methyl-2-(tributylstannyl)indol
Formel: C₂₁H₃₅NSn
MW: 420,21 g/mol
MDL Number: MFCD09265483
CAS-Nummer: 157427-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR