2-Methyl-5-(tributylstannyl)furan

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51229G1 AS51229G5
ACTIAS51229G1EA 243 CHF
ACTIAS51229G1 ACTIAS51229G5
2-Methyl-5-(tributylstannyl)furan
2-Methyl-5-(tributylstannyl)furan
Formel: C₁₇H₃₂OSn
MDL Number: MFCD01319081
CAS-Nummer: 118486-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR