2,5-Bis(tributylstannyl)furan

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51702G1 AS51702G5
ACTIAS51702G1EA 0 CHF
ACTIAS51702G1 ACTIAS51702G5
2,5-Bis(tributylstannyl)furan
2,5-Bis(tributylstannyl)furan
Formel: C₂₈H₅₆OSn₂
MDL Number: MFCD09833740
CAS-Nummer: 193361-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR