Tributyl(2-furyl)stannan

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51074G1 AS51074G5
ACTIAS51074G1EA 0 CHF
ACTIAS51074G1 ACTIAS51074G5
Tributyl(2-furyl)stannan
2-(Tributylstannyl)furan
Formel: C₁₆H₃₀OSn
MW: 357,10 g/mol
MDL Number: MFCD00192512
CAS-Nummer: 118486-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR