2-(Tributylstannyl)benzo[b]thiophen
Lieferant: ACTIVATE SCIENTIFIC
|
Gefahr
|
AS51614G1
AS51614G5
ACTIAS51614G1EA
0
CHF
ACTIAS51614G1
ACTIAS51614G5
2-(Tributylstannyl)benzo[b]thiophen
2-(Tributylstannyl)benzo[b]thiophen
Formel:
C₂₀H₃₂SSn MW: 423.22 g/mol |
MDL Number:
MFCD01318936 CAS-Nummer: 148961-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...