3,6-Dihydro-4-(tributylstannyl)-1(2H)-pyridincarbonsäure-tert-butylester

Lieferant: ACTIVATE SCIENTIFIC
Gefahr

AS51766G1 AS51766G5
ACTIAS51766G1EA 0 CHF
ACTIAS51766G1 ACTIAS51766G5
3,6-Dihydro-4-(tributylstannyl)-1(2H)-pyridincarbonsäure-tert-butylester
3,6-Dihydro-4-(tributylstannyl)-1(2H)-pyridincarbonsäure-tert-butylester
Formel: C₁₆H₃₀O₂Sn
MDL Number: MFCD18325165
CAS-Nummer: 208465-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR