4-(Morpholino)phenylboronsäurepinakolester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine

AS21039G1 AS21039G25 AS21039G5
ACTIAS21039G1EA 0 CHF
ACTIAS21039G1 ACTIAS21039G25 ACTIAS21039G5
4-(Morpholino)phenylboronsäurepinakolester
4-(Morpholino)phenylboronsäurepinakolester
Formel: C₁₆H₂₄BNO₃
MW: 289.18 g/mol
MDL Number: MFCD04112544
CAS-Nummer: 568577-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR