4-(4-Morpholinylcarbonyl)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-(4-Morpholinylcarbonyl)benzeneboronic acid, (4-(Morpholine-4-carbonyl)phenyl)boronic acid

AS51468G1 AS51468G5
ACTIAS51468G1EA 0 CHF
ACTIAS51468G1 ACTIAS51468G5
4-(4-Morpholinylcarbonyl)phenylboronsäure
4-(4-Morpholinylcarbonyl)phenylboronsäure
Formel: C₁₁H₁₄BNO₄
MW: 235.05 g/mol
MDL Number: MFCD03411952
CAS-Nummer: 389621-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR